Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8388 4-in-1, Type R, Class 3/2, Thermal High Visibility Jacket Kit

Catalog No. 19820993 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-993 Lime Large
19-820-991 Lime Small
19-820-992 Lime Medium
19-820-994 Lime X-Large
19-820-995 Lime 2X-Large
19-820-996 Lime 3X-Large
19-820-997 Lime 4X-Large
19-820-998 Lime 5X-Large
19-820-999 Orange Small
19-821-000 Orange Medium
19-821-003 Orange Large
19-821-004 Orange X-Large
19-821-005 Orange 2X-Large
19-821-006 Orange 3X-Large
19-910-100 Orange 4X-Large
19-910-101 Orange 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-993 Supplier Ergodyne™ Supplier No. 25534
Add to Cart
Add to Cart
This item is not returnable. View return policy

GloWear™ 8388 4-in-1 Thermal High Visibility Jacket Kit includes an outershell jacket and a thermal jacket.

  • Weatherproof 300D oxford polyester outer shell
  • Breathable PU coating
  • Inner jacket with 100 g 3M™ Thinsulate™ lining
  • Inner jacket zips out and sleeves zip off to create Class 2 thermal vest
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Black panels hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Two external front pockets with snap flaps
  • External zipper chest pocket
  • Cellphone chest pocket
  • Clear ID badge holder
  • Dual mic tabs for clipping radios
  • YKK #5 zippers

Specifications

Size Large
Color Lime
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.