Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8910BK Class E Black Bottom Hi-Vis Pants

Catalog No. 19820758 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small, Medium
Large, X-Large
2X-Large, 3X-Large
4X-Large, 5X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-758 2X-Large, 3X-Large
19-820-756 Small, Medium
19-820-757 Large, X-Large
19-820-759 4X-Large, 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-820-758 Supplier Ergodyne™ Supplier No. 23957
Add to Cart
Add to Cart
This item is not returnable. View return policy

Class E Black Bottom Mesh Pants feature black high wear areas to hide dirt and keep worker looking clean.

  • ANSI-compliant polyester mesh (3.3 oz.)
  • 2 in. ANSI-compliant reflective
  • Drawstring waist
  • Wide leg openings with hook and loop closures
  • One right leg cargo pocket
  • Black accents on bottom of leg high wear areas

Specifications

Size 2X-Large, 3X-Large
Product Type Hi-vis Mesh Pants
Material Polyester
Color Yellow
No. of Pockets 1
For Use With (Application) Road Construction, Utility Crew
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.