Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8915 Class E Rain Pants

Catalog No. 19820863 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Orange
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-863 Yellow X-Large
19-820-852 Orange Small
19-820-853 Orange Medium
19-820-854 Orange Large
19-820-855 Orange X-Large
19-820-856 Orange 2X-Large
19-820-857 Orange 3X-Large
19-820-858 Orange 4X-Large
19-820-859 Orange 5X-Large
19-820-860 Yellow Small
19-820-861 Yellow Medium
19-820-862 Yellow -
19-820-864 Yellow 2X-Large
19-820-865 Yellow -
19-820-866 Yellow 4X-Large
19-820-867 Yellow 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-863 Supplier Ergodyne™ Supplier No. 24425
Add to Cart
Add to Cart
This item is not returnable. View return policy

GloWear™ 8915 Class E Rain Pants designed to provide breathable, waterproof comfort.

  • Weatherproof 300D oxford outer shell
  • Breathable PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket

Specifications

Size X-Large
Material Polyurethane
Color Yellow
Waist Style Elastic with drawstring
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.