Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8915BK Class E Black Bottom Rain Pants

Catalog No. 19820943 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
4X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-943 X-Large
19-820-940 Small
19-820-941 Medium
19-820-944 2X-Large
19-820-946 -
19-820-947 4X-Large
19-820-948 -
19-820-942 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-820-943 Supplier Ergodyne™ Supplier No. 25425
Add to Cart
Add to Cart
This item is not returnable. View return policy

Class E Black Bottom Rain Pants offer breathable, waterproof, comfort with black accents to hide dirt.

  • Weatherproof 300D oxford outer shell
  • Breathable PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket
  • Black accents on bottom of leg

Specifications

Size X-Large
Color Yellow
Waist Style Elastic with drawstring
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.