Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8918BK Class E Black Bottom Rain Bibs

Catalog No. 19820911 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-911 3X-Large
19-820-901 Small
19-820-902 Medium
19-820-903 Large
19-820-904 X-Large
19-820-908 2X-Large
19-820-916 4X-Large
19-820-917 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-820-911 Supplier Ergodyne™ Supplier No. 25017
Add to Cart
Add to Cart
This item is not returnable. View return policy

Class E Black Bottom Rain Bibs provide breathable, waterproof, comfort in a bib style with black accents to hide dirt.

  • Weatherproof 300D oxford outer shell
  • Breathable PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Adjustable elastic suspenders
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket
  • Black accents on bottom of leg

Specifications

Size 3X-Large
Material Polyurethane
Color Yellow
Waist Style Adjustable elastic suspenders
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.