Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8925 Class E Thermal Pants

Catalog No. 19820869 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Orange
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-869 Orange Medium
19-820-868 Orange Small
19-820-870 Orange Large
19-820-872 Orange 2X-Large
19-820-873 - 3X-Large
19-820-874 - 4X-Large
19-820-875 Orange 5X-Large
19-820-876 Yellow Small
19-820-877 - Medium
19-820-878 Yellow Large
19-820-879 Yellow X-Large
19-820-880 Yellow 2X-Large
19-820-881 Yellow 3X-Large
19-820-882 Yellow 4X-Large
19-820-883 Yellow 5X-Large
19-820-871 Orange X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-869 Supplier Ergodyne™ Supplier No. 24443
Add to Cart
Add to Cart
This item is not returnable. View return policy

Class E Thermal Pants feature breathable 3M™ Thinsulate™ warmth and waterproof comfort.

  • Weatherproof 100% polyester outer shell
  • Breathable PU coating
  • 80 g 3M™ Thinsulate™ lining
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket

Specifications

Size Medium
Color Orange
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.