Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8925 Class E Thermal Pants

Catalog No. 19820874 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Color:
Orange
Yellow
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-820-874 4X-Large -
19-820-868 Small Orange
19-820-869 Medium Orange
19-820-870 Large Orange
19-820-872 2X-Large Orange
19-820-873 3X-Large -
19-820-875 5X-Large Orange
19-820-876 Small Yellow
19-820-877 Medium -
19-820-878 Large Yellow
19-820-879 X-Large Yellow
19-820-880 2X-Large Yellow
19-820-881 3X-Large Yellow
19-820-882 4X-Large Yellow
19-820-883 5X-Large Yellow
19-820-871 X-Large Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-874 Supplier Ergodyne™ Supplier No. 24448
Add to Cart
Add to Cart
This item is not returnable. View return policy

Class E Thermal Pants feature breathable 3M™ Thinsulate™ warmth and waterproof comfort.

  • Weatherproof 100% polyester outer shell
  • Breathable PU coating
  • 80 g 3M™ Thinsulate™ lining
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket

Specifications

Size 4X-Large
Material Polyester
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.