Learn More
Description
- ANSI Class E Complaint for low light conditions
- Made with tough 300D polyester oxford to withstand wear and tear
- PU transparent coating provides wind and water-resistant protection
- Scuff-resistant knee patches for extra durability
- Black accents in high-wear areas to hide dirt and grime
- Adjustable straps and buckle release for comfort and fit
- 1 zippered chest pocket and 2 side hand pockets for storage
- Boot zippers for easy on and off without removing footwear
Specifications
Specifications
| Size | 2X-Large |
| Material | Polyester |
| Color | Yellow |
| For Use With (Application) | Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.