Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8928 Class E Hi-Vis Insulated Bibs

Catalog No. 19820984 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-984 3X-Large
19-820-979 Small
19-820-980 Medium
19-820-981 Large
19-820-982 X-Large
19-820-983 2X-Large
19-820-985 4X-Large
19-820-986 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-820-984 Supplier Ergodyne™ Supplier No. 25527
Add to Cart
Add to Cart
This item is not returnable. View return policy

GloWear™ 8928 Hi-Vis Insulated Bibs provide conspicuity, warmth, and durability for the toughest conditions.

  • ANSI Class E Complaint for low light conditions
  • Made with tough 300D polyester oxford to withstand wear and tear
  • PU transparent coating provides wind and water-resistant protection
  • Scuff-resistant knee patches for extra durability
  • Black accents in high-wear areas to hide dirt and grime
  • Adjustable straps and buckle release for comfort and fit
  • 1 zippered chest pocket and 2 side hand pockets for storage
  • Boot zippers for easy on and off without removing footwear

Specifications

Size 3X-Large
Material Polyester
Color Yellow
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.