Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ Class 3 Zip-Up Hi-Vis Hooded Sweatshirt with Black Front

Catalog No. 19820618 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-618 4X-Large
19-820-613 Medium
19-820-614 Large
19-820-615 X-Large
19-820-616 2X-Large
19-820-617 3X-Large
19-820-619 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-820-618 Supplier Ergodyne™ Supplier No. 21848
This item is not returnable. View return policy

Designed with high-quality 100% polyester polar fleece to provide visibility and warmth on chilly days

  • Safety sweatshirt has two slash-style pockets for additional hand warmth and storage
  • bottom front and wrist sections of the sweatshirt are black to prevent dirt from showing
  • Two inch ANSI-compliant silver reflective tape on torso, shoulders and sleeves for effective visibility as night falls
  • Zipper front closure, drawstring hood and elastic knit waistband and cuffs keep the heat in for all-day warmth
  • Ideal for jobsites in high-traffic areas like road construction or anywhere that requires high visibility clothing or for activities such as biking, snowmobiling, snow shoeing, and ice fishing

Specifications

Size 4X-Large
Material Polyester Polar Fleece
Color Hi-vis Lime
Certifications/Compliance ANSI/ISEA 107-2015 Type R, Class 3 compliant
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction, Railway Workers, Warehouse/Distribution
Includes Drawstring hood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.