Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8281BK Type R Class 2 Black Front Performance Long Sleeve T-Shirt

Catalog No. 19820683 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Hi-vis Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-683 Hi-vis Lime 2X-Large
19-820-679 Hi-vis Lime Small
19-820-680 Hi-vis Lime Medium
19-820-681 Hi-vis Lime Large
19-820-682 Hi-vis Lime X-Large
19-820-684 Hi-vis Lime 3X-Large
19-820-687 Orange Small
19-820-688 Orange Medium
19-820-689 Orange Large
19-820-690 Orange X-Large
19-820-691 Orange 2X-Large
19-820-692 Orange 3X-Large
19-820-693 Orange 4X-Large
19-820-694 Orange 5X-Large
19-820-685 Hi-vis Lime 4X-Large
19-820-686 Hi-vis Lime 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-683 Supplier Ergodyne™ Supplier No. 22636
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight birdseye knit paired with 3M™ Scotchlite™ Comfort Trim™

Specifications

Size 2X-Large
Color Hi-vis Lime
Certifications/Compliance ANSI-compliant polyester breathable knit
For Use With (Application) Road Construction, Utility Crew, Survey Crew, Construction, Railway Workers, Baggage Handlers, Warehouse/Distribution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.