Learn More
Ergodyne™ GloWear™ 8281BK Type R Class 2 Black Front Performance Long Sleeve T-Shirt
Shop All Ergodyne ProductsSpecifications
Specifications
| Size | 2X-Large |
| Color | Orange |
| Certifications/Compliance | ANSI-compliant polyester breathable knit |
| For Use With (Application) | Road Construction, Utility Crew, Survey Crew, Construction, Railway Workers, Baggage Handlers, Warehouse/Distribution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.