Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8289BK Type R Class 2 Black Front T-Shirt

Catalog No. 19820662 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Hi-vis Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-662 Orange 5X-Large
19-820-647 Hi-vis Lime Small
19-820-648 Hi-vis Lime Medium
19-820-649 Hi-vis Lime Large
19-820-650 Hi-vis Lime X-Large
19-820-651 Hi-vis Lime 2X-Large
19-820-652 Hi-vis Lime 3X-Large
19-820-653 Hi-vis Lime 4X-Large
19-820-654 Hi-vis Lime 5X-Large
19-820-655 Orange Small
19-820-656 Orange Medium
19-820-657 Orange Large
19-820-658 Orange X-Large
19-820-659 Orange 2X-Large
19-820-660 Orange 3X-Large
19-820-661 Orange 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-662 Supplier Ergodyne™ Supplier No. 22519
This item is not returnable. View return policy

Lightweight birdseye knit t-shirt with black front to keep high-wear area looking clean.

Specifications

Size 5X-Large
Color Orange
No. of Pockets 1
Certifications/Compliance ANSI-compliant polyester breathable knit
For Use With (Application) Road Construction, Utility Crew, Survey Crew, Construction, Railway Workers, Baggage Handlers, Warehouse/Distribution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.