Learn More
Specifications
Specifications
| Certifications/Compliance | ANSI-compliant polyester mesh |
| Color | Lime |
| Size | 2X-Large/3X-Large |
| For Use With (Application) | Road Construction, Utility Crew, Survey Crew, Construction, Railway Workers, Baggage Handlers, Emergency Response, Warehousing/Distribution |
| Material | Polyester |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.