Learn More
CPC Scientific H-His-Asp-Glu-Phe-Glu-Arg-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-Gly-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GLUC008A
Encompass
GLP, synthesized by posttranslational processing of proglucagon in the intestine and pancreas, plays an important role in metabolic homeostasis. It is capable of converting intestinal epithelial cells into insulin-producing cells, thus serving as a new putative therapeutic compound for the treatment of diabetes mellitus. ONE-LETTER SEQUENCE: HDEFERHAEGTFTSDVSSYLEGQAAKEFIAWLVKGRGMOLECULAR FORMULA: C186H275N51O59MOLECULAR WEIGHT:4169.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [87805-34-3], RESEARCH AREA: DiabetesREFERENCES: G.I. Bell et al., Nature, 304, 368 (1983)
Catalog No. 50-210-9373
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.