Learn More
Enzo Life Sciences Glucagon like peptide 1 (human) (7-36) amide (100µg)
SDP
Supplier:  Enzo Life Sciences BMLGP14380100
Encompass_Preferred
A predominant cleaved form of pro-glucagon expressed in the intestine. GLP-1 [7-36]amide is promptly released from the gut after oral ingestion of a meal where it slows gastric emptying and exerts a powerful insulinotropic action leading to elevated serum insulin levels. GLP-1 [7-36] amide promotes satiety and reduces food intake in patients with diabetes mellitus type 2 and has been suggested to have therapeutic potential in the treatment of NIDDM. It has been reported that human pancreatic endocrine tumors contain molecular forms of GLP-1 which resemble those expressed in the small intestine, rather than in their tissue of origin. Alternative name: GLP 1 (7-36) amide. Solubility: Soluble in water at 1mg/mL. Long Term Storage: -20°C.
Catalog No. 50-201-0907
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.