Learn More
Alpco Ltd Total GLP-1 ELISA (7-36 and 9-36)
SDP
Supplier:  Alpco Ltd 43GPTHUE01
Encompass_Preferred
The Total GLP-1 ELISA (7-36 and 9-36) is a highly sensitive ELISA used exclusively for the quantitative determination of bioactive glucagon like peptide-1 (7-36) and (9-36) levels in human EDTA-plasma.
Superior analytical sensitivity
0.6 pmol/L
Broad dynamic range
2.1-54 pmol/L
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-922-298
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.