Learn More
Cayman Chemical GLP-17-13humnmouser 10mg
SDP
Supplier:  Cayman Chemical 4125510 MG
Encompass_Preferred
An ionizable cationic lipid (pKa 6.512) administration of SARS-CoV-2 Omicron variant mRNA in YK009-containing LNPs induces SARS-CoV-2 RBD-binding IgG in mice with a boost in antigen-specific IgG titers 10 days after a second dose
Catalog No. 50-294-5024
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.