Learn More
Bachem GLP-2 (rat)
SDP
Supplier:  Bachem 4034866.0500
Encompass_Preferred
GLP-2 (rat) , H-His-Ala-Asp-Gly-Ser-Phe-Ser-Asp-Glu-Met-Asn-Thr-Ile-Leu-Asp-Asn-Leu-Ala-Thr-Arg-Asp-Phe-Ile-Asn-Trp-Leu-Ile-Gln-Thr-Lys-Ile-Thr-Asp-OH, 195262-56-7, C166H256N44O56S, Mr 3796.19, 0.5mg. Preproglucagon (146-178) (rat), Glucagon-Like Peptide 2 (rat), Proglucagon (126-158) (rat), Glucagon-Like Peptide II, rat. GLP-2 administration to mice or rats promotes stimulation of crypt cell proliferation and inhibition of enterocyte apoptosis resulting in hyperplasia of the small bowel villous epithelium. It also exerts trophic effects in animal models of both small and large bowel injury such as experimental small bowel resection or chemically induced colitis. In addition to stimulation of epithelial proliferation, GLP-2 also acutely regulates gastric emptying and exerts rapid metabolic effects promoting stimulation of intestinal hexose transport. The actions of GLP-2 are transduced via the GLP-2 receptor, a member of the glucagon-secretin G protein-coupled receptor superfamily.
Catalog No. 50-194-6691
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.