Learn More
Alpco Ltd GLP-2 ELISA, 96 Wells, 25 L, Plasma, Serum
SDP
Supplier:  Alpco Ltd 48GP2HUE01.1
Encompass_Preferred
The GLP-2 ELISA is designed for determination of GLP-2 in Plasma and Serum samples. The assay design of the GLP-2 ELISA is based on a competitive enzyme immunoassay format using a highly specific antibody to human GLP-2 and a biotin – avidin affinity system. The GLP-2 ELISA includes all components (96 well Ab coated plate, labeled antigen, substrate, standards and dilution and wash buffer) necessary to complete the GLP-2 ELISA. 
Catalog No. 50-922-558
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.