Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLP-1 Monoclonal Antibody (4)
GREENER_CHOICE

Catalog No. ABA330402
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABA330402 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABA330402 Supplier Invitrogen™ Supplier No. ABS0330402
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABS 033-04-02 detects the amidated N-terminus of GLP-1(7-37) and GLP-1(7-36) in human samples. ABS 033-04-02 has been successfully used in ELISA procedures. ABS 033-04-02 binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36) amide and shows < 0.2% cross-reactivity with GLP-1(1-37), GLP-1(9-36) amide, glucagon, human GIP and exendin-4. ABS 033-04-02 cross-reacts approximately 1% with human GLP-2. Not recommended for sandwich ELISA applications. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

GCG is also known as GLP1, or Glucagon. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis.It is mapped to 2q36-2q37. GLP1, also known as 7-37 for the codons of the preproglucagon molecule which encode it, renders pancreatic beta-cells 'glucose-competent' and may be useful in the treatment of noninsulin-dependent diabetes mellitus. Also, GLP1 is a potent insulin secretagogue.It plays a major role in the enteroinsular axis, accounting, for example, for the finding that plasma insulin levels accompanying oral intake of glucose are greater than those observed when glucose is given intravenously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLP-1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 4
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene GCG
Gene Accession No. P01275
Gene Alias GCG; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; glucagon; Glucagon like peptide 1; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; Incretin hormone; MGC138331; OXM; OXY; Oxyntomodulin; PPG; preproglucagon
Gene Symbols GCG
Host Species Mouse
Immunogen Synthetic GLP-1 (7-17).
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2641
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.