Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLP1R Recombinant Rabbit Monoclonal Antibody (HL2297)
GREENER_CHOICE

Catalog No. FEMA550480 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550480 Supplier Invitrogen™ Supplier No. MA550480
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

GLP1R Recombinant Monoclonal Antibody for IHC (P)

Glucagon-like peptide-1(9-36)amide and GLP-1(9-37) are the forms of GLP-1 that result from the rapid degradation of the active forms of the peptide (GLP-1(7-36)amide and GLP-1(7-37)) by the enzyme dipeptidyl peptidase-IV (DPP-IV, also known as CD26 or adenosine deaminase binding protein). GLP-1 is a peptide hormone of the glucagon family, produced by the L cells of the intestinal mucosa from the same prohormone as glucagon. The active forms are potent stimulators of glucose-dependent insulin secretion. The sequence of GLP-1 is fully conserved in all mammalian species examined so far.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLP1R
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2297
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Glp1r
Gene Accession No. O35659
Gene Alias Glip; GLP-1; GLP1 receptor; GLP-1 receptor; GLP1R; GLP-1R; GLP-1-R; GLP1Rc; GLP-I receptor; Glpr; glucagon like peptide 1 receptor; glucagon-like peptide 1 receptor; glucagon-like peptide-1 receptor; LOW QUALITY PROTEIN: glucagon-like peptide 1 receptor; glucagon-like peptide 1 receptor; pancreatic beta cell receptor for the gluco-incretin hormone glucagon-like peptide 1; RATGL1RCP; seven transmembrane helix receptor
Gene Symbols Glp1r
Host Species Rabbit
Immunogen Synthetic peptide of human GLP1R.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14652
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.