Learn More
Apexbio Technology LLC GLPG0634, 5mg. Cas: 1206161-97-8 MFCD: MFCD20527867. JAK1 inhibitor
SDP
Supplier:  Apexbio Technology LLC B11305
Encompass_Preferred
GLPG0634 is a selective inhibitor of JAK1 with IC50 value of 10nM [1].GLPG0634 is a small molecule inhibitor of JAK1. It shows potent inhibitory activity against both JAK1 and JAK2 with IC50 values of 10nM and 28nM, respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5207
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.