Learn More
Creative Biomart Dulaglutide, Recombinant GLP1
Supplier:  Creative Biomart THP0117
Encompass_Preferred
The product is a novel glucagon-like peptide-1 agonist (GLP-1) biologic drug consisting of a dipeptidyl peptidase-IV-protected GLP-1 analogue covalently linked to a human IgG4-Fc heavy chain by a small peptide linker.
Catalog No. 50-208-4168
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.