Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Gluconolactone, 99%

Catalog No. AC271050010
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271050010 1 kg Glass bottle
AC271052500 250 g Glass bottle
AC271050051 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC271050010 Supplier Thermo Scientific Chemicals Supplier No. 271050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 90-80-2 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-80-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00006647
InChI Key PHOQVHQSTUBQQK-UHFFFAOYNA-N
Synonym gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone
PubChem CID 7027
ChEBI CHEBI:16217
SMILES OCC1OC(=O)C(O)C(O)C1O

Specifications

Melting Point 151.0°C to 155.0°C
Color White
Density 0.6000g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 kg
Beilstein 18, 405
Specific Gravity 0.6
Merck Index 15, 4493
Specific Rotation + 65.00
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: 1g/100 g alcohol,insoluble in ether
IUPAC Name (3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
Formula Weight 178.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material δ-Gluconolactone
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement MOISTURE SENSITIVE
EINECSNumber 202-016-5
RTECSNumber LZ5184000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.