Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glucose, 2-amino-2-deoxy- | 3416-24-8 | MFCD09025572 | ≥98.0% | 179.17 | C6H13NO5 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB112510MG

Encompass_Preferred

Glucosamine (D-glucosamine) is an amino sugar and precursor in the biosynthesis of glycosylated proteins and lipids, used as a reagent in metabolic pathway and cell biology research.

  • Amino sugar precursor in glycosylation and metabolic pathway research.
  • High purity (≥98.0% by NMR) suitable for analytical and biological studies.
  • Molecular weight 179.17 g/mol; formula C6H13NO5.
  • Appearance: white to off-white solid, for laboratory handling and storage.
  • Available in small research pack sizes (e.g., 10 mg).

Catalog No. 50-002-97631


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.