Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucosamine Sulfate Potassium Chloride, Dietary Supplement, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602940 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
12 kg
25 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-940 12 kg Poly Pail
18-602-941 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-940 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G129612KG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

G1296, 1296149-08-0, (C6H14NO5)2SO4·2KCl

Spectrum™ Chemical Glucosamine Sulfate Potassium Chloride, Dietary Supplement, USP slows deterioration of cartilage, relieves osteoarthritis (OA) pain and improves joint mobility. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1296149-08-0

Specifications

CAS Max % 100%
Ignition Residue 26.5-31%
Loss on Drying 1%
Packaging Poly Pail
Quantity 12 kg
Specific Rotation 47.0° to +53.0
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.