Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 5%, Innovating Science™
SCI_ED

Catalog No. S031089 Shop All Aldon Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031089 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S031089 Supplier Aldon Corporation Supplier No. IS16016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides a convenient solution for preparing a 5% concentration.

Chemical Identifiers

Molecular Weight (g/mol) 180.16 g/mol

Specifications

Boiling Point Approx. 100°C
Quantity 25 g
Percent Purity 100%
Grade Not Available
Concentration or Composition (by Analyte or Components) 5%
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.