Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Glucose-6-phosphate Dehydrogenase

Catalog No. ICN15118701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 U
1kU
500 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15118701 1kU
02151187-1 100 U
02151187-5 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15118701 Supplier MP Biomedicals™ Supplier No. 0215118701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose-6-phosphate Dehydrogenase is an enzyme in pentose phosphate pathway.

  • Glucose-6-phosphate Dehydrogenase (G-6-PDH) was used as a model to test the effect of seed protein fractions on enzyme protection during dehydration.
  • G-6-PDH has been utilized in assays for nicotinamide adenine dinucleotide and tissue pyridine nucleotides.
TRUSTED_SUSTAINABILITY

Specifications

CAS 9001-40-5
Content And Storage Store at -20°C
Quantity 1kU
Product Type Glucose-6-Phosphate Dehydrogenase
Form Solid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.