Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Glucose Dry Powder Reagents

Catalog No. 23666292 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
10 x 100mL (3.4 oz.)
1000 mL
5 x 600 mL
Form:
Dry Powder
Liquid
Formulation:
Glucose Oxidase >15U/mL, Peroxidase (horseradish) 800U/L, 4-Aminoantipyrine >15U/mL, Phosphate Buffer pH 7.5 ± 0.1, Sodium p-Hydroxybenzoate, non-reactive Stabilizers and fillers, Sodium Azide 0.1%
Glucose Oxidase (microbial) 12000U/L, Peroxidase (horseradish) >1000U/L,4-AAP >0.3mM, Phenol 4mM, Buffer pH 7.4 ± 0.1, non-reactive Stabilizers
pH:
7.3
7.4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Formulation pH
23-666-292 1000 mL Dry Powder Glucose Oxidase >15U/mL, Peroxidase (horseradish) 800U/L, 4-Aminoantipyrine >15U/mL, Phosphate Buffer pH 7.5 ± 0.1, Sodium p-Hydroxybenzoate, non-reactive Stabilizers and fillers, Sodium Azide 0.1% 7.4
23-666-275 5 x 600 mL Liquid Glucose Oxidase (microbial) 12000U/L, Peroxidase (horseradish) >1000U/L,4-AAP >0.3mM, Phenol 4mM, Buffer pH 7.4 ± 0.1, non-reactive Stabilizers 7.3
23-666-276 10 x 100mL (3.4 oz.) Liquid Glucose Oxidase (microbial) 12000U/L, Peroxidase (horseradish) >1000U/L,4-AAP >0.3mM, Phenol 4mM, Buffer pH 7.4 ± 0.1, non-reactive Stabilizers 7.3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-292 Supplier Pointe Scientific Supplier No. G75195L

missing translation for 'validMainframeMsg'

Includes

  • Glucose Oxidase >15u/ml; Peroxidase (horseradish) 800U/L; 4-
  • Aminoantipyrine 0.38mM; Phosphate Buffer; pH 7.5±0.1; Sodium p-Hydroxybenzoate 5mM; non-reactive Stabilizers and fillers; Sodium Azide 0.1%

Quantify glucose levels in serum samples

  • Method: Oxidase/Trinder, Endpoint
  • Format: Dry powder
  • Wavelength: 500nm
  • Linearity: 500mg/dL
  • Expected Values: 70 to 105mg/dL
  • Reconstituted stability: 30 days at 2 to 8°C

For the quantitative determination of glucose in serum

Specifications

Certifications/Compliance CE Marked
Type Glucose (Oxidase) Reagent Set
Formulation Glucose Oxidase >15U/mL, Peroxidase (horseradish) 800U/L, 4-Aminoantipyrine >15U/mL, Phosphate Buffer pH 7.5 ± 0.1, Sodium p-Hydroxybenzoate, non-reactive Stabilizers and fillers, Sodium Azide 0.1%
For Use With (Application) Quantitative determination of Glucose in Serum, in-vitro diagnostic use only
pH 7.4
Quantity 1000 mL
Form Dry Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.