Learn More
Invitrogen™ Glucose Oxidase Monoclonal Antibody (GO13-103.6.2)

Description
MA1-10603 detects Glucose Oxidase from Aspergillus niger samples. MA1-10603 has been successfully used in ELISA applications. The MA1-10603 immunogen is purified glucose oxidase from Aspergillus niger.
Specifications
Specifications
| Antigen | Glucose Oxidase |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | GO13-103.6.2 |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | aspergillus niger glucose oxidase; GOx |
| Host Species | Mouse |
| Immunogen | Purified glucose oxidase from Aspergillus niger. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.