Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Glucose Uptake Assay Kit, Fluorometric, Quantitative, 100 Tests

Catalog No. 502583615
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3615 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3615 Supplier Abcam Supplier No. AB136956100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides a quantitative, 2-hour measurement of D-glucose levels in cell samples with 0.05 nmol/well sensitivity.

  • Detection method: Fluorescent
  • Sensitivity: 0.05 nmol/well
  • Assay time: 2 hr
  • Sample types: Adherent cells, Suspension cells
  • Results type: Quantitative
  • Instrument compatibility: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule D-Glucose
Substrate Properties Chemical Substrate
Substrate D-Glucose
Run Time 2 hr.
No. of Reactions 100 Tests
Detection Location In-Solution Detection
For Use With (Equipment) Microplate Reader
Format Kit
Detection Method Fluorescence
Sample Type Adherent cells, Suspension cells
Quantity 100 Tests
For Use With (Application) Glucose uptake measurement
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.