Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluR2/GluR3 Recombinant Rabbit Monoclonal Antibody (4K7G5)
GREENER_CHOICE

Catalog No. PIMA542796
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA542796 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIMA542796

Supplier: Invitrogen™ MA542796

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Mouse brain, Rat brain. Immunogen sequence: KNKWWYDKGE CGSGGGDSKE KTSALSLSNV AGVFYILVGG LGLAMLVALI EFCYKSRAEA KRMKVAKNAQ NINPSSSQNS QNFATYKEGY NVYGIESVKI.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate)-sensitive glutamate receptors, and is subject to RNA editing (AGA->GGA; R->G). Alternative splicing at this locus results in different isoforms, which may vary in their signal transduction properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR2/GluR3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 4K7G5
Concentration 1.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gria2
Gene Accession No. P19491, P19492, P23819, Q9Z2W9
Gene Alias 2900064I19Rik; AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA2; GluA3; GLUR2; GluR-2; GLUR3; GluR-3; Gluralpha3; GLURB; gluR-B; GLURC; GLUR-C; GluR-K2; GluR-K3; glutamate ionotropic receptor AMPA type subunit 2; glutamate ionotropic receptor AMPA type subunit 3; glutamate receptor; glutamate receptor 2; Glutamate receptor 3; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor C; glutamate receptor channel alpha3 subunit; glutamate receptor ionotropic, AMPA 2; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 3; glutamate receptor, ionotropic, AMPA2 (alpha 2); glutamate receptor, ionotropic, AMPA3 (alpha 3); GPRC1C; GRIA2; GRIA3; GRM3; HBGR2; Kiaa4184; MGR3; MRX94
Gene Symbols Gria2, Gria3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 784-883 of human GluR2+GluR3 (P42262).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14800, 29627, 29628, 53623
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.