Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluR2 Monoclonal Antibody (6C4)
GREENER_CHOICE

Catalog No. 320300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-030-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-030-0 Supplier Invitrogen™ Supplier No. 320300
  • $479.00
    $359.65 / Each of 1

    Save $119.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

GluR2 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5 -methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain. The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6C4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Gria2
Gene Accession No. P19491, P23819, P42262
Gene Alias AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; GluA2; Glur2; GluR-2; GLURB; GluR-B; GluR-K2; glutamate ionotropic receptor AMPA type subunit 2; glutamate receptor; glutamate receptor 2; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA2 (alpha 2); GRIA2; HBGR2; OTTHUMP00000165324; OTTHUMP00000220942; OTTHUMP00000220943
Gene Symbols Gria2
Host Species Mouse
Immunogen Fusion protein containing amino acids 175-430 from the N-terminal region of GluR2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14800, 2891, 29627
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.