Learn More
Enzo Life Sciences GLUT1 monoclonal antibody (2475) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7620100
Encompass_Preferred
Clone: 2475. Isotype: IgG2Bκ. Immunogen: A portion of amino acids 203-305 from the human protein was used as the immunogen for this GLUT1 antibody. Species reactivity: Human. Specificity: Recognizes a protein of 45-55kDa, which is identified as GLUT1. Applications: ELISA, Flow Cytometry, IF, IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-3301
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.