Learn More
Invitrogen™ GLUT1 Monoclonal Antibody (C3)

Description
GLUT1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | GLUT1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | C3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | SLC2A1 |
| Gene Accession No. | P11166 |
| Gene Alias | choreoathetosis/spasticity, episodic (paroxysmal choreoathetosis/spasticity); CSE; DYT17; DYT18; DYT9; EIG12; glucose transporter glut1; glucose transporter type 1, erythrocyte/brain; glucose transporter type I; GLUT; Glut1; Glut-1; GLUT1DS; GLUTB; GT1; GTG1; Gtg3; HepG2 glucose transporter; HTLVR; human T-cell leukemia virus (I and II) receptor; MGC141895; MGC141896; PED; RATGTG1; receptor for HTLV-1 and HTLV-2; SDCHCN; Slc2a1; solute carrier family 2 (facilitated glucose transporter), member 1; Solute carrier family 2 a 1 (facilitated glucose transporter) brain; solute carrier family 2 member 1; solute carrier family 2, facilitated glucose transporter member 1; solute carrier family 2, member 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.