Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLUT14 Polyclonal Antibody, Invitrogen™

Catalog No. p-7226345
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121934 20 μL
PIPA5121933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121934 Supplier Invitrogen™ Supplier No. PA5121934
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC2A14 (Solute Carrier Family 2 Member 14) is a Protein Coding gene. Diseases associated with SLC2A14 include Hypoascorbemia and Amyotrophic Lateral Sclerosis 1. Among its related pathways are Metabolism and Transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds. Gene Ontology (GO) annotations related to this gene include transmembrane transporter activity and glucose transmembrane transporter activity. An important paralog of this gene is SLC2A3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SLC2A14
Gene Accession No. Q8TDB8
Gene Alias GLUT-14; GTR14
Gene Symbols SLC2A14
Host Species Rabbit
Immunogen Recombinant GLUT14 (Met1-Val105) expressed in E.coli.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144195
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.