Learn More
AdipoGen GLUT2 rat pAb IN119
SDP
Supplier:  AdipoGen AG25B0043C050
Encompass_Preferred
Polyclonal Antibody. Recognizes rat GLUT2. Applications IHC, IP, WB. Source Rabbit. GLUT2 belongs to the family of glucose transporters GLUTs, encoded by the SLC2A genes, which comprises 14 isoforms. GLUT2 has a uniquely low affinity for glucose Km ~ 17mmol/l, and can also use mannose, galactose and fructose as low-affinity substrates. However, it has a high affinity for glucosamine Km ~ 0.8mmol/l. GLUT2 is expressed in liver, intestine, kidney and pancreatic islet beta cells, as well as in the central nervous system, in neurons, astrocytes and tanycytes. Physiological studies of genetically modified mice have revealed a role for GLUT2 in several regulatory mechanisms. GLUT2 is also the major glucose transporter in the plasma membrane of hepatocytes. In pancreatic beta cells, GLUT2 is required for glucose-stimulated insulin secretion.
Catalog No. 50-256-3179
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.