Learn More
GLUT3 Recombinant Rabbit Monoclonal Antibody (6T3F2)

Description
GLUT3 Recombinant Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | GLUT3 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 6T3F2 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC2A3 |
| Gene Accession No. | P11169, P32037, Q07647 |
| Gene Alias | AA408729; AL023014; AL024341; AU040424; C78366; FLJ90380; glucose transporter 3; glucose transporter type 3, brain; Glut3; Glut-3; Slc2a3; solute carrier family 2 (facilitated glucose transporter), member 3; Solute carrier family 2 A3 (neuron glucose transporter); solute carrier family 2 member 3; solute carrier family 2, facilitated glucose transporter member 3; solute carrier family 2, member 2; solute carrier family 2, member 3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.