Learn More
Description
This antibody was generated against the internal region of human GLUT3 and may cross-react with GLUT14.20403-1-AP antibody detects the bands around 48-60 kDa in SDS-PAGE.
Specifications
Specifications
| Antigen | GLUT3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC2A3 |
| Gene Accession No. | P11169, P32037 |
| Gene Alias | GLUT 3, GLUT3, SLC2A3 |
| Gene Symbols | SLC2A3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
