Learn More
Invitrogen™ GLUT9 Polyclonal Antibody

Description
Cross-reactivity with human samples is very low. Suggested positive control: human kidney membrane protein.
Specifications
Specifications
| Antigen | GLUT9 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% sodium azide |
| Gene | SLC2A9 |
| Gene Accession No. | Q3T9X0, Q9NRM0 |
| Gene Alias | facilitated glucose transporter, type 9; glucose transporter type 9; GLUT9; GLUT-9; GLUTX; human glucose transporter-like protein-9; Slc2a9; SLC2a9A; SLC2A9B; solute carrier family 2 (facilitated glucose transporter), member 9; solute carrier family 2 member 9; solute carrier family 2, facilitated glucose transporter member 9; UAQTL2; Urate transporter; urate voltage-driven efflux transporter 1; URATv1 |
| Gene Symbols | SLC2A9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.