Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutahione oxidized, For HPLC analysis, 98%, MP Biomedicals™

Catalog No. ICN15119305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15119305 5 g
ICN1511932 250 mg
ICN15119301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15119305 Supplier MP Biomedicals, Inc Supplier No. 0215119305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27025-41-8
Molecular Formula C20H32N6O12S2
Molecular Weight (g/mol) 612.63
MDL Number MFCD00150701
InChI Key YPZRWBKMTBYPTK-IOKZUGQQNA-N
Synonym oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
PubChem CID 65359
ChEBI CHEBI:17858
SMILES N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

Specifications

Color White
Quantity 5 g
IUPAC Name 2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid
Formula Weight 612.64
Percent Purity 0.98
Physical Form Lyophilized Powder
Chemical Name or Material Glutahione oxidized, For HPLC analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.