Learn More
MOLECULAR DEPOT LLC RF Serum 20000-24999 - 1 mL
Supplier:  MOLECULAR DEPOT LLC B2010861F
Encompass_Preferred
High Rheumatoid Factor Positive Serum is human serum containing high levels (20000-24999 IU/mL) of Rheumatoid Factor antibodies (RF) Titer has been measured with the RF Latex method No preservatives have been added All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units For in vitro research use only
Catalog No. 50-000-13332