Learn More
Bioassay Systems QuantiChrom™ Glutamate Dehydrogenase Assay Kit. For quantitative determination of glutamate dehydrogenase activity.
SDP
Catalog No. 50489206 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of glutamate dehydrogenase activity. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.4 to 80 U/L for 30 min reaction. Detection Limit of 0.1 U/L for 120 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Serum, plasma, cell, tissue, agriculture etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.1 U/L.
Catalog No. 50-489-206 Supplier Bioassay Systems Supplier No. DGLDH100
For quantitative determination of glutamate dehydrogenase activity. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.4 to 80 U/L for 30 min reaction. Detection Limit of 0.1 U/L for 120 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Serum, plasma, cell, tissue, agriculture etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.1 U/L.