Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ GlutaMAX™ Supplement
GREENER_CHOICE

Catalog No. 35050061
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
20 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-050-061 100 mL
35-050-079 20 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-050-061 Supplier Gibco™ Supplier No. 35050061
Add to Cart
Edge
Add to Cart

GlutaMAX Supplement is an L-alanyl-L-glutamine dipeptide alternative to L-glutamine. It exhibits increased stability over L-glutamine leading to improved cellular health.

Gibco GlutaMAX Supplement is an L-alanyl-L-glutamine dipeptide alternative to L-glutamine that exhibits increased stability, leading to improved cell health. L-Glutamine is an amino acid required in cell culture for the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, and other amino acids, as well as in protein synthesis and glucose production. GlutaMAX Supplement is suitable for adherent and suspension culture of mammalian cells, with no adaptation required. It is formulated as a 200 mM L-alanyl-L-glutamine dipeptide in 0.85% NaCl.

Compared to L-glutamine, GlutaMAX Supplement:

  • Minimizes toxic ammonia build-up
  • Improves cell viability and growth
  • Remains stable across a wide range of temperatures

Minimizes toxic ammonia build-up

Unlike L-glutamine, GlutaMAX Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.

Improves cell viability and growth

Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX Supplement that is not possible with L-glutamine.

Remains stable across a wide range of temperatures

GlutaMAX Supplement is room-temperature stable and ready to use. It retains full functionality for 24 months at room temperature, 2 to 8°C, or -5 to -30°C for convenient storage and handling. GlutaMAX Supplement is even stable at 37°C for a week or more.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 15°C to 30°C
Alternate storage conditions: -5°C to -20°C and 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 100X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type GlutaMax
Sterility Sterile-filtered
For Use With (Application) Clinical Research, Translational Research
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 430 to 490 mOsm/kg
pH 5 to 6
Product Line Gibco, GlutaMAX
Quantity 100 mL
Shipping Condition Room Temperature
Source Animal Origin-free
Show More Show Less
Why is GlutaMAX Supplement (Cat. Nos. 35050061, 35050079) recommended over L-glutamine when reconstituting Geltrex Reduced Growth Factor Basement Membrane Matrix?

GlutaMAX Supplement (Cat. No. 35050061, 35050079) is highly recommended to reconstitute Geltrex Reduced Growth Factor Basement Membrane Matrix because it offers higher stability than L-glutamine. The L-glutamine in Geltrex matrix comes in the form of the dipeptide L-alanyl-L-glutamine, which degrades slower than L-glutamine in cell culture. While GlutaMAX supplement is highly recommended, it is not required for Geltrex matrix–based culture systems.

Does Dynamis AGT Medium (Cat. Nos. A2617501, A2617502, A2617503, A2617504) require supplementation for use?

When using Dynamis AGT Medium (Cat. Nos. A2617501, A2617502, A2617503, A2617504) for culturing a CHO cell line other than the glutamine synthetase (GS) gene expression system (GS-CHO expression system), the addition of 2-8 mM L-glutamine or a similar amount of GlutaMAX Supplement (Cat. No. 35050079, 35050061) is recommended.

Note: We recommend that you supplement with glucose as needed so that glucose levels don’t fall below 2g/L in culture (see the product User Guide for more details).

Do my cells need to adapt to GlutaMAX Supplement (Cat. No. 35050061, 35050079)?

Some cell lines may require an adaptation period, and the culture may lag in growth until it becomes accustomed to GlutaMAX Supplement (Cat. No. 3050061, 35050079). For serum-free media it is best to add L-glutamine along with GlutaMAX supplement. We don’t recommend that you use GlutaMAX supplement with insect cells such as Sf9.

What is the difference between GlutaMAX Supplement (Cat. No. 35050061), CTS GlutaMAX-I Supplement (Cat. No. A1286001), and CTS GlutaMAX Supplement (Cat. No. A4737001)?

The formulation and manufacturing process are the same for all three products. The main difference is in the “intended use” statement. CTS GlutaMAX-I Supplement (Cat. No. A1286001) and CTS GlutaMAX Supplement (Cat. No. A4737001) have an intended use statement of “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals”. GlutaMAX Supplement (Cat. No. 35050061) has an intended use statement of “For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals”. The catalog number to use will depend on the specific application.

Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?

In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.

What is the difference between Gibco GlutaMAX and glutamine supplements?

GlutaMAX Supplement is an improved cell culture supplement that can be used as a direct substitute for L-glutamine in your cell culture medium. Please go here to read more about the properties of GlutaMAX supplement (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/media-supplements/glutamax-media/glutamax-vs-glutamine.html).


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.