Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glutaminase C (GAC) Monoclonal Antibody (GT3211)
GREENER_CHOICE

Catalog No. PIMA531538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531538 Supplier Invitrogen™ Supplier No. MA531538
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Predicted reactivity: Bovine (100%). Positive Control: MCF-7, MDA-MB-231. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutaminase C (GAC)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT3211
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GLS
Gene Accession No. D3Z7P3, O94925
Gene Alias 6330442B14; AAD20; AI314027; B230365M23Rik; GAC; GAM; GLS; Gls1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; RATGLUT
Gene Symbols GLS
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Glutaminase C (GAC). The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14660, 2744
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.