Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Glutaminase (GLS1) Inhibitor Screening Kit (Fluorometric), 100 Tests

Catalog No. 502205383
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-5383 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-5383 Supplier Abcam Supplier No. AB283389100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Antibody, suitable for Enzyme activity assay.

  • Applications: Enzyme activity assay

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen GLS2
Conjugate Unconjugated
Gene GLS2
Gene Symbols GLS2
Quantity 100 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Target Species Non-species specific
Product Type Antibody (GLS2)
Detection Method Fluorescent
Shipping Condition Blue Ice
Target Gene GLS2
Target Entry GLS2
Target Molecule GLS2
Technique Enzyme activity assay
Validated Application abrv Enzyme activity assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.