Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glutaminase Recombinant Rabbit Monoclonal Antibody (6H5L15)

Catalog No. 701965 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-196-5 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-196-5 Supplier Invitrogen™ Supplier No. 701965
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Rabbit, Rat and Mouse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Glutaminase is a phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. It is primarily expressed in the brain and kidney and plays an essential role in generating energy for metabolism, synthesizing neurotransmitter glutamate, and maintaing acid-based balance in the kidney. There are alternate splicing results in multiple transcript variants in the Glutaminase gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutaminase
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6H5L15
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene GLS
Gene Accession No. D3Z7P3, O94925, P13264
Gene Alias 6330442B14; AAD20; AI314027; B230365M23Rik; DKFZp686O15119; FLJ10358; GA; GAC; GAM; GLS; GLS1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; PAG; phosphate-activated glutaminase; RATGLUT
Gene Symbols GLS
Host Species Rabbit
Immunogen Protein corresponding to Human GLS (aa 251-533).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14660, 24398, 2744
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.