Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ STING/TMEM173 Antibody Blocking Peptide
SDP

Catalog No. NBP224683SP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224683SP 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224683SP Supplier Novus Biologicals™ Supplier No. NBP224683PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

The STING/TMEM173 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 340061
Species Human
Common Name STING/TMEM173
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation diH2O
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP2-24683. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias anti-STING, endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, human STING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, mouse STING, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173
Gene Symbol TMEM173
Molecular Weight (g/mol) M.W. (theoretical): 42 kDa
Quantity 0.05 mg
Research Category Cancer, Immunology, Innate Immunity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.