Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific γ-Glutamyl Transferase Reagents for Hitachi™ Analyzers

Catalog No. 23666263 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
3 x 289 R2mL
3 x 410 R1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-263 3 x 289 R2mL
23-666-262 3 x 410 R1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-666-263 Supplier Pointe Scientific Supplier No. HG459R2

missing translation for 'validMainframeMsg'

Includes

Reagent 1 and Reagent 2 when combined: Tris buffer (pH 8.1±0.1) 89mM; Glycylglycine 126mM; GLUPA-C 3.2mM; Sodium azide 0.095%; stabilizer

Quantify γ-GT easily with system specific reagents

  • Method: Modified Szasz, Kinetic
  • Format: Liquid (ready to use)
  • Linearity: 800 U/L
  • Expected Values: 6 to 54 U/L

Applications:

For the quantitative kinetic determination of γ-glutamyl transferase (γ-GT) activity in serum on Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type gamma-Glutamyl Transferase R2 Kit
For Use With (Application) Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum
Quantity 3 x 289 R2mL
Form Liquid

For use with in vitro diagnostics only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.